Benzeneruthenium(II) chloride dimer 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dichloro(benzene)ruthenium(II) dimer

AC36748-0010 AC36748-0050
TS36748-0010EA 163 USD
TS36748-0010 TS36748-0050
Benzeneruthenium(II) chloride dimer 97%
Benzeneruthenium(II) chloride dimer
Formula: C₁₂H₁₂Cl₄Ru₂
Melting Pt: 249 °C
Storage Temperature: Ambient
MDL Number: MFCD00064686
CAS Number: 37366-09-9

Order Now

Specification Test Results

Appearance Red to brown Powder or crystals
Infrared spectrum Conforms
Ruthenium (Ru) 39.2 to 41.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR