Allylpalladium(II) chloride dimer 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Di-mu-chlorobis[(eta-allyl)palladium(II)], Allylpalladium(II) chloride, Allylpalladium chloride dimer

AC20683-0050 AC20683-5000
TS20683-0050EA 920 USD
TS20683-0050 TS20683-5000
Allylpalladium(II) chloride dimer 98%
Allylpalladium(II) chloride dimer
Formula: C₆H₁₀Cl₂Pd₂
Storage Temperature: Freezer
MDL Number: MFCD00044874
CAS Number: 12012-95-2

Order Now

Specification Test Results

Appearance Yellow to yellow-green Crystalline powder or crystals
Assay ≥56.7 % (Pd)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR