1,3-Divinyltetramethyldisiloxane 97%

Supplier: Thermo Scientific Chemicals
Warning

AC24921-0100 AC24921-0500 AC24921-2500
TS24921-0100EA 39 USD
TS24921-0100 TS24921-0500 TS24921-2500
1,3-Divinyltetramethyldisiloxane 97%
1,3-Divinyltetramethyldisiloxane
Formula: C₈H₁₈OSi₂
Boiling Pt: 139 °C (760 mmHg)
Melting Pt: –99.7 °C
Density: 0.813 g/cm³
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00014933
CAS Number: 2627-95-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4130 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR