1,2-Bis(2-iodoethoxy)ethane 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethylene Glycol Bis(2-iodoethyl) Ether

AC39169-0050 AC39169-0250
TS39169-0050EA 46 USD
TS39169-0050 TS39169-0250
1,2-Bis(2-iodoethoxy)ethane 97% stabilized
1,2-Bis(2-iodoethoxy)ethane
Formula: C₆H₁₂I₂O₂
MW: 369.97 g/mol
Density: 2.028 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075008
CAS Number: 36839-55-1

Order Now

Specification Test Results

Appearance Clear yellow to orange-red Liquid
Infrared spectrum Conforms
GC ≥96.0 %
Stabilizer copper

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR