(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dichloro(pentamethylcyclopentadienyl)iridium(III) Dimer, Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, [Ir(Cp-)Cl2]2, (Pentamethylcyclopentadienyl)iridium (III) chloride dimer

AC37795-0010 AC37795-0050 AC37795-2500
TS37795-0010EA 391 USD
TS37795-0010 TS37795-0050 TS37795-2500
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer 99%
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer
Formula: C₂₀H₃₀Cl₄Ir₂
MW: 796.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075435
CAS Number: 12354-84-6

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Infrared spectrum Conforms
Assay ≥98.5 % (ex Ir)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR