(Acetylacetonato)(norbornadiene)rhodium(I)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (Acetylacetonato)[bicyclo[2.2.1]hepta-2,5-diene]rhodium(I), (Norbornadiene)rhodium(I) 2,4-pentanedionate

AC38256-1000 AC38256-5000
TS38256-1000EA 90 USD
TS38256-1000 TS38256-5000
(Acetylacetonato)(norbornadiene)rhodium(I)
(Acetylacetonato)(norbornadiene)rhodium(I)
Formula: C₁₂H₁₅O₂Rh
MW: 294.16 g/mol
Melting Pt: 140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075051
CAS Number: 32354-50-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Rhodium (Rh) ≥34.7 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR