(3-Cyanopropyl)dimethylchlorosilane 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chloro(3-cyanopropyl)dimethylsilane

AC31338-0100 AC31338-0500
TS31338-0100EA 73 USD
TS31338-0100 TS31338-0500
(3-Cyanopropyl)dimethylchlorosilane 95%
(3-Cyanopropyl)dimethylchlorosilane
Formula: C₆H₁₂ClNSi
MW: 161.70 g/mol
Boiling Pt: 230…235 °C (760 mmHg)
Density: 0.99 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00001969
CAS Number: 18156-15-5
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to straw yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4470 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR