(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate 99%

Supplier: Thermo Scientific Chemicals
Warning

AC36358-1000 AC36358-5000
TS36358-1000EA 110 USD
TS36358-1000 TS36358-5000
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate 99%
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate
Formula: C₃₁H₅₀F₆IrNP₂
Melting Pt: 175 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00075097
CAS Number: 64536-78-3

Order Now

Specification Test Results

Appearance Orange Powder
Infrared spectrum Conforms
Carbon (C) 45.6 to 47.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR