4-Bromopiperidine hydrobromide 98%

Supplier: Thermo Scientific Chemicals
Warning

AC36831-0010 AC36831-0100
TS36831-0010EA 40 USD
TS36831-0010 TS36831-0100
4-Bromopiperidine hydrobromide 98%
4-Bromopiperidine hydrobromide
Formula: C₅H₁₁Br₂N
Melting Pt: 199…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00191858
CAS Number: 54288-70-9

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 191 to 196 °C
Titration Argentometric 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR