Bis(dibenzylideneacetone)palladium

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(dibenzylideneacetone)palladium(0)

AC29197-0010 AC29197-0050
TS29197-0010EA 83 USD
TS29197-0010 TS29197-0050
Bis(dibenzylideneacetone)palladium
Bis(dibenzylideneacetone)palladium
Formula: C₃₄H₂₈O₂Pd
MW: 575 g/mol
Melting Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00051942
CAS Number: 32005-36-0

Order Now

Specification Test Results

Appearance Red-brown to black Powder
Assay 16 to 21 % (Pd)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR