2-(Pyridin-4-yl)acetic acid hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Pyridylacetic acid hydrochloride

AC29475-0010 AC29475-0050
TS29475-0010EA 31 USD
TS29475-0010 TS29475-0050
2-(Pyridin-4-yl)acetic acid hydrochloride 98%
2-(Pyridin-4-yl)acetic acid hydrochloride
Formula: C₇H₈ClNO₂
MW: 173.60 g/mol
Melting Pt: 139…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00012827
CAS Number: 6622-91-9

Order Now

Specification Test Results

Appearance White to cream Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR