Bis(acetonitrile)palladium(II) dichloride 99%

Supplier: Thermo Scientific Chemicals
Warning

AC20945-0050 AC20945-5000
TS20945-0050EA 678 USD
TS20945-0050 TS20945-5000
Bis(acetonitrile)palladium(II) dichloride 99%
Formula: C₄H₆Cl₂N₂Pd
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013122
CAS Number: 14592-56-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Orange to rust brown Crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR