1,3-Bis(diphenylphosphino)propanenickel(II) chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: [1,3-Bis(diphenylphosphino)propane]nickel (II) chloride, Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II), 1,3-Bis(diphenylphosphino)propane nickel(II)chloride, Dichloro[1,3-bis(diphenylphosphino)propane]nickel(II), [1,3-Bis(diphenylphosphino)propane]dichloronickel(II), [1,3-Bis(diphenylphosphino)propane]nickel(II) dichloride

AC29159-0010 AC29159-0050 AC29159-0250
TS29159-0010EA 39 USD
TS29159-0010 TS29159-0050 TS29159-0250
1,3-Bis(diphenylphosphino)propanenickel(II) chloride 99%
1,3-Bis(diphenylphosphino)propanenickel(II) chloride
Formula: C₂₇H₂₆Cl₂NiP₂
MW: 542.042 g/mol
Melting Pt: 213 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00015318
CAS Number: 15629-92-2

Order Now

Specification Test Results

Appearance Red to red-brown Powder or crystals
Infrared spectrum Conforms
Elemental Analysis ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR