2-(Pyridin-3-yl)acetic acid hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyridyl-3-acetic acid hydrochloride, 3-Pyridylacetic acid hydrochloride

AC13196-0025 AC13196-0250
TS13196-0025EA 57 USD
TS13196-0025 TS13196-0250
2-(Pyridin-3-yl)acetic acid hydrochloride 98%
2-(Pyridin-3-yl)acetic acid hydrochloride
Formula: C₇H₈ClNO₂
MW: 173.60 g/mol
Melting Pt: 160…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00012819
CAS Number: 6419-36-9

Order Now

Specification Test Results

Appearance White to light yellow to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
HPLC ≥96.0 %
Specific optical rotation -35° to -45° (20°C, 589 nm) (c=1, chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR