Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tris(dibenzylideneacetone)dipalladium(0) chloroform adduct, Tris(dibenylideneacetone)dipalladium-chloroform, Tris(dibenzylideneacetone)dipalladium-chloroform adduct
AC36934-0010
AC36934-0050
TS36934-0010EA
111
USD
TS36934-0010
TS36934-0050
Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform 97%
Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform
Formula:
C₅₁H₄₂O₃Pd₂·CHCl₃ MW: 1035.11 g/mol Melting Pt: 131…135 °C Storage Temperature: Freezer |
MDL Number:
MFCD00075479 CAS Number: 52522-40-4 |
Specification Test Results
Appearance | Purple to black Crystalline powder |
Assay | ≥96.0 % (ex Pd) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...