Sodium ammonium hydrogen phosphate tetrahydrate 99% for analysis

Supplier: Thermo Scientific Chemicals

Synonyms: XSodium ammonium hydrogen phosphate tetrahydrate, Sodium ammonium phosphate, Ammonium sodium phosphate dibasic tetrahydrate, Sodium ammonium phosphate tetrahydrate, Ammonium sodium phosphate tetrahydrate

AC22264-0010 AC22264-0025 AC22264-2500
TS22264-0010EA 77 USD
TS22264-0010 TS22264-0025 TS22264-2500
Sodium ammonium hydrogen phosphate tetrahydrate 99% for analysis
Sodium ammonium hydrogen phosphate tetrahydrate
Formula: NaNH₄HPO₄·4H₂O
MW: 209.07 g/mol
Melting Pt: 80 °C
Density: 1.54 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066970
CAS Number: 7783-13-3

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder or crystals
Titration with HCl ≥98.5 %
Chloride (Cl) ≤30 ppm
Nitrate (NO3) ≤20 ppm
Sulfate (SO4) ≤100 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤10 ppm
Insoluble matter ≤0.01 % (in NH4OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR