Potassium ethyltrifluoroborate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potassium ethyltrifluoroboranuide

AC44488-0010 AC44488-0050
TS44488-0010EA 105 USD
TS44488-0010 TS44488-0050
Potassium ethyltrifluoroborate 97%
Potassium ethyltrifluoroborate
Formula: C₂H₅BF₃K
MW: 135.9656 g/mol
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD04112713
CAS Number: 44248-07-9

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
Melting point ≥260 °C
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR