Potassium wolframate 99.95% -100 mesh for trace metal analysis

Supplier: Thermo Scientific Chemicals
Warning

AC44833-0050 AC44833-0250
TS44833-0050EA 29 USD
TS44833-0050 TS44833-0250
Potassium wolframate 99.95% -100 mesh for trace metal analysis
Potassium wolframate
Formula: K₂O₄W
Melting Pt: 921 °C
Storage Temperature: Ambient
MDL Number: MFCD00049668
CAS Number: 7790-60-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Total trace metal impurities ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR