4-(Trimethylsilyl)-3-butyn-1-ol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Trimethylsilyl-3-butyn-1-ol

AC38132-0050 AC38132-0250
TS38132-0050EA 128 USD
TS38132-0050 TS38132-0250
4-(Trimethylsilyl)-3-butyn-1-ol 98%
4-(Trimethylsilyl)-3-butyn-1-ol
Formula: C₇H₁₄OSi
MW: 142.27 g/mol
Boiling Pt: 97…98 °C (24 mmHg)
Density: 0.854 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00190217
CAS Number: 2117-12-6
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4540 to 1.4560 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR