1-Ethoxy-1-(tributylstannyl)ethene 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tributyl(1-ethoxyvinyl)tin, Tri-n-butyl(1-ethoxyvinyl)tin, 1-Ethoxyvinyltri-n-butyltin

AC37771-0010 AC37771-0050 AC37771-0250
TS37771-0010EA 43 USD
TS37771-0010 TS37771-0050 TS37771-0250
1-Ethoxy-1-(tributylstannyl)ethene 95%
1-Ethoxy-1-(tributylstannyl)ethene
Formula: C₁₆H₃₄OSn
Boiling Pt: 85…86 °C (.1 mmHg)
Density: 1.069 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00010240
CAS Number: 97674-02-7
UN: 2788
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4770 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR