4-Acetylphenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone, 4-Acetylphenylboronic acid, pinacol ester, 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone

AC44443-0010 AC44443-0050
TS44443-0010EA 78 USD
TS44443-0010 TS44443-0050
4-Acetylphenylboronic acid pinacol ester 97%
4-Acetylphenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₃
Boiling Pt: 80 °C (.1Torr)
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD05863922
CAS Number: 171364-81-1

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR