Tetrakis(trimethylsilyl)silane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
TMSS
AC29390-0010
AC29390-0050
TS29390-0010EA
43
USD
TS29390-0010
TS29390-0050
Tetrakis(trimethylsilyl)silane 97%
Tetrakis(trimethylsilyl)silane
Formula:
C₁₂H₃₆Si₅ MW: 320.842 g/mol Melting Pt: >300 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00054859 CAS Number: 4098-98-0 |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...