Calcium perchlorate tetrahydrate, pure

Supplier: Thermo Scientific Chemicals
Danger

AC39063-1000 AC39063-5000
TS39063-1000EA 42 USD
TS39063-1000 TS39063-5000
Calcium perchlorate tetrahydrate, pure
Calcium perchlorate tetrahydrate
Formula: CaClO₄·4H₂O
Storage Temperature: Ambient
MDL Number: MFCD00191895
CAS Number: 15627-86-8
UN: 1455
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Complexometric 12.2 to 13.5 % (Calcium)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR