2-(Trimethylsilyl)ethoxymethyl chloride 90% tech. stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC21902-0050
AC21902-0250
AC21902-1000
TS21902-0050EA
167
USD
TS21902-0050
TS21902-0250
TS21902-1000
2-(Trimethylsilyl)ethoxymethyl chloride 90% tech. stabilized
2-(Trimethylsilyl)ethoxymethyl chloride
Formula:
C₆H₁₅ClOSi MW: 166.72 g/mol Boiling Pt: 57…59 °C (8 mmHg) Density: 0.94 g/cm³ Flash Pt: 46 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009919 CAS Number: 76513-69-4 EINECS: 278-483-4 UN: 2920 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4340 to 1.4380 (20°C, 589 nm) |
GC | ≥88 % |
Stabilizer | ≤200 ppm Diisopropylethylamine |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...