Cerium(III) carbonate hydrate 99.9% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

AC37940-0100 AC37940-0500 AC37940-2500
TS37940-0100EA 22 USD
TS37940-0100 TS37940-0500 TS37940-2500
Cerium(III) carbonate hydrate 99.9% (trace metals basis)
Cerium (III) carbonate hydrate
Formula: C₃Ce₂O₉·H₂O
MW: 478.274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149635
CAS Number: 54451-25-1
EINECS: 208-655-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Total trace metal impurities ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR