Potassium selenocyanate 99%

Supplier: Thermo Scientific Chemicals
Danger

AC20198-0100 AC20198-0500 AC20198-2500
TS20198-0100EA 99 USD
TS20198-0100 TS20198-0500 TS20198-2500
Potassium selenocyanate 99%
Potassium selenocyanate
Formula: CKNSe
Melting Pt: 152…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00011366
CAS Number: 3425-46-5
EINECS: 222-320-1
UN: 3283
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or needles
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR