2-Aminophenylboronic acid hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

AC44531-0010 AC44531-0050
TS44531-0010EA 111 USD
TS44531-0010 TS44531-0050
2-Aminophenylboronic acid hydrochloride 97%
2-Aminophenylboronic acid hydrochloride
Formula: C₆H₉BClNO₂
MW: 173.4052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02258096
CAS Number: 863753-30-4

Order Now

Specification Test Results

Appearance White to light yellow Powder or solid
Infrared spectrum Conforms
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR