Rhodium(II) acetate dimer dihydrate ca 43% Rh

Supplier: Thermo Scientific Chemicals
Warning

AC19761-0010 AC19761-2500
TS19761-0010EA 1152 USD
TS19761-0010 TS19761-2500
Rhodium(II) acetate dimer dihydrate ca 43% Rh
Rhodium (II) acetate dimer dihydrate
Formula: C₈H₁₂O₈Rh₂·2H₂O
Storage Temperature: Ambient
CAS Number: 29998-99-0
EINECS: 240-084-8

Order Now

Specification Test Results

Appearance Green Powder
Infrared spectrum Conforms
Assay 43 % (Rh)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR