Potassium vinyltrifluoroborate 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium trifluoro(vinyl)borate

AC38770-0010 AC38770-0050
TS38770-0010EA 48 USD
TS38770-0010 TS38770-0050
Potassium vinyltrifluoroborate 95%
Potassium vinyltrifluoroborate
Formula: C₂H₃BF₃K
MW: 133.95 g/mol
Melting Pt: 240…242 °C
Storage Temperature: Ambient
MDL Number: MFCD02093335
CAS Number: 13682-77-4
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Assay ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR