1-Octylmagnesium chloride 1.4 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC42803-1000 AC42803-8000
TS42803-1000EA 82 USD
TS42803-1000 TS42803-8000
1-Octylmagnesium chloride 1.4 M in THF, AcroSeal™
1-Octylmagnesium chloride
Formula: C₈H₁₇ClMg
Flash Pt: –10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000476
CAS Number: 38841-98-4
UN: 2924
ADR: 3 II

Order Now

Specification Test Results

Appearance Gray-green to brown Solution
Titration Grignard 1.2 to 1.6 M
Additional info is possible)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR