Dimethylethoxysilane 94%

Supplier: Thermo Scientific Chemicals
Danger

AC39200-0050 AC39200-0250 AC39200-1000
TS39200-0050EA 28 USD
TS39200-0050 TS39200-0250 TS39200-1000
Dimethylethoxysilane 94%
Dimethylethoxysilane
Formula: C₄H₁₂OSi
MW: 104.23 g/mol
Flash Pt: –5 °C
Storage Temperature: Ambient
MDL Number: MFCD00026750
CAS Number: 14857-34-2
EINECS: 238-921-7
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3670 to 1.3700 (20°C, 589 nm)
GC ≥92 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR