Hexaammineruthenium trichloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexaammineruthenium(III) chloride, Hexaammineruthenium (III) chloride

AC36334-0010 AC36334-0050 AC36334-2500
TS36334-0010EA 150 USD
TS36334-0010 TS36334-0050 TS36334-2500
Hexaammineruthenium trichloride 98%
Hexaammineruthenium trichloride
Formula: H₁₈Cl₃N₆Ru
MW: 309.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011478
CAS Number: 14282-91-8
EINECS: 238-176-8

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Elemental Analysis ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR