Dibismuth trisulfate 99%, pure
Supplier: Thermo Scientific Chemicals
Synonyms:
Bismuth (III) sulphate, Bismuth (III) sulfate
AC45385-0250
AC45385-1000
TS45385-0250EA
50
USD
TS45385-0250
TS45385-1000
Dibismuth trisulfate 99%, pure
Dibismuth trisulphate
Formula:
Bi₂O₁₂S₃ MW: 706.13 Melting Pt: 405 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00036140 CAS Number: 7787-68-0 |
Specification Test Results
Appearance | White Powder |
Assay | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...