Bis(tri-tert-butylphosphine)palladium(0) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC36350-0010
AC36350-1000
TS36350-0010EA
314
USD
TS36350-0010
TS36350-1000
Bis(tri-tert-butylphosphine)palladium(0) 98%
Bis(tri-tert-butylphosphine)palladium(0)
Formula:
C₂₄H₅₄P₂Pd Melting Pt: 258…272 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03094580 CAS Number: 53199-31-8 |
Specification Test Results
Appearance | Off-white to orange-brown Powder or crystals |
Infrared spectrum | Conforms |
Assay | ≥97.5 % (ex Pd) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...