Bis(1,5-cyclooctadiene)nickel(0)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Bis(1,5-cyclooctadiene)nickel(0) (Wax encapsulated) (ca. 0.05mmol/capsule), Ni(COD)2
AC22397-0010
AC22397-0050
AC22397-0250
TS22397-0010EA
92
USD
TS22397-0010
TS22397-0050
TS22397-0250
Bis(1,5-cyclooctadiene)nickel(0)
Bis(1,5-cyclooctadiene)nickel (0)
Formula:
C₁₆H₂₄Ni MW: 275.06 g/mol Melting Pt: 60 °C Storage Temperature: Freezer |
MDL Number:
MFCD00058902 CAS Number: 1295-35-8 EINECS: 215-072-0 UN: 1325 ADR: 4.1 II |
Specification Test Results
Appearance | Yellow to orange Powder or crystals |
Nickel (Ni) | 20.0 to 22.6 % (with dimethylglyoxime) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...