Chloro(1,5-cyclooctadiene)iridium(I) dimer

Supplier: Thermo Scientific Chemicals
Warning

AC36396-0010 AC36396-0050 AC36396-2500
TS36396-0010EA 465 USD
TS36396-0010 TS36396-0050 TS36396-2500
Chloro(1,5-cyclooctadiene)iridium(I) dimer
Chloro(1,5-cyclooctadiene)iridium (I) dimer
Formula: C₁₆H₂₄Cl₂Ir₂
Melting Pt: 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012414
CAS Number: 12112-67-3
EINECS: 235-170-7

Order Now

Specification Test Results

Appearance Orange to red Powder
Infrared spectrum Conforms
Iridium (Ir) ≥56.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR