Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

Supplier: Thermo Scientific Chemicals
Danger

AC37565-0010 AC37565-2500
TS37565-0010EA 661 USD
TS37565-0010 TS37565-2500
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Bis(1,5-cyclooctadiene)rhodium (I) tetrafluoroborate
Formula: C₁₆H₂₄BF₄Rh
Melting Pt: 210 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00075045
CAS Number: 35138-22-8
EINECS: 460-220-1
UN: 2925
ADR: 4.1,II

Order Now

Specification Test Results

Appearance Red Powder
Infrared spectrum Conforms
Rhodium (Rh) ≥24.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR