Nickel(II) hydroxide for analysis

Supplier: Thermo Scientific Chemicals
Danger

AC27502-0010 AC27502-2500
TS27502-0010EA 185 USD
TS27502-0010 TS27502-2500
Nickel(II) hydroxide for analysis
Nickel(II) hydroxide
Formula: H₂NiO₂
MW: 92.71 g/mol
Melting Pt: 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011140
CAS Number: 12054-48-7
EINECS: 235-008-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Green Crystalline powder
Titration Complexometric 61.0 to 65.0 % (Ni)
Cobalt (Co) ≤8000 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤200 ppm
Manganese (Mn) ≤50 ppm
Sulfur (S) ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR