Potassium tetraoxalate dihydrate 99%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Oxalic acid, potassium salt dihydrate, Potassium trihydrogen dioxalate dihydrate
AC35211-0010
AC35211-1000
AC35211-5000
TS35211-0010EA
536
USD
TS35211-0010
TS35211-1000
TS35211-5000
Potassium tetraoxalate dihydrate 99%, Extra Pure
Potassium tetraoxalate dihydrate
Formula:
C₄H₃KO₈·2H₂O MW: 254.19 (218.16 anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150443 CAS Number: 6100-20-5 EINECS: 204-874-6 |
Specification Test Results
Appearance | Colorless to white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | 12 % to 16 % (1 g, 105°C) |
Titration with NaOH | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...