Tetrakis(dimethylamido)titanium(IV) 99.99% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC35356-0050
AC35356-0250
TS35356-0050EA
126
USD
TS35356-0050
TS35356-0250
Tetrakis(dimethylamido)titanium(IV) 99.99% (trace metals basis)
Tetrakis(dimethylamino)titanium (TDMAT)
Formula:
C₈H₂₄N₄Ti Flash Pt: –30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014861 CAS Number: 3275-24-9 EINECS: 221-904-3 UN: 3129 ADR: 4.3,I |
Specification Test Results
Appearance | Clear yellow to orange Liquid |
Infrared spectrum | Conforms |
Total trace metal impurities | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...