Tris(trimethylsilyl)phosphite 92%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(trimethylsilyl)phosphite

AC42852-0050 AC42852-0250 AC42852-1000
TS42852-0050EA 88 USD
TS42852-0050 TS42852-0250 TS42852-1000
Tris(trimethylsilyl)phosphite 92%
Tris(trimethylsilyl) phosphite
Formula: C₉H₂₇O₃PSi₃
MW: 298.54 g/mol
Boiling Pt: 78…81 °C (8.3 mmHg)
Density: 0.893 g/cm³
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00015588
CAS Number: 1795-31-9

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4080 to 1.4110 (20°C, 589 nm)
GC ≥90 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR