O-tert-Butylhydroxylamine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: O-(tert-Butyl)hydroxylamine hydrochloride

AC39968-0010 AC39968-0050
TS39968-0010EA 115 USD
TS39968-0010 TS39968-0050
O-tert-Butylhydroxylamine hydrochloride 99%
O-tert-Butylhydroxylamine hydrochloride
Formula: C₄H₁₂ClNO
MW: 125.5972 g/mol
Melting Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00043272
CAS Number: 39684-28-1
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Titration Argentometric 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR