Sodium bis(2-methoxyethoxy)aluminium hydride ∼3.5 M in toluene 70% (w/w), AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium dihydrobis(2-methoxyethoxy)aluminate, SBAH, Sodium bis(2-methoxyethoxy)aluminum dihydride

AC43091-1000 AC43091-8000
TS43091-1000EA 45 USD
TS43091-1000 TS43091-8000
Sodium bis(2-methoxyethoxy)aluminium hydride ∼3.5 M in toluene 70% (w/w), AcroSeal™
Sodium bis(2-methoxyethoxy)aluminium hydride
Formula: C₆H₁₆AlNaO₄
MW: 202.16 g/mol
Boiling Pt: 110 °C (760 mmHg)
Density: 1.036 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00011631
CAS Number: 22722-98-1
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear to slightly hazy colorless to amber Viscous liquid
Assay 68.0 to 72.0 %
Melting point 50 to 54 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR