Sodium bis(2-methoxyethoxy)aluminium hydride ∼3.5 M in toluene 70% (w/w)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium dihydrobis(2-methoxyethoxy)aluminate, SBAH, Sodium bis(2-methoxyethoxy)aluminum dihydride
AC18790-0010
AC18790-0500
AC18790-2500
TS18790-0010EA
186
USD
TS18790-0010
TS18790-0500
TS18790-2500
Sodium bis(2-methoxyethoxy)aluminium hydride ∼3.5 M in toluene 70% (w/w)
Sodium bis(2-methoxyethoxy)aluminium hydride
Formula:
C₆H₁₆AlNaO₄ Density: 1.036 g/cm³ Flash Pt: 4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011631 CAS Number: 22722-98-1 EINECS: 245-178-2 UN: 3399 ADR: 4.3,I |
Specification Test Results
Appearance | Clear to slightly hazy colorless to amber Viscous liquid |
Assay | 68.0 to 72.0 % |
Comments | Liquid |
D-Enrichment | ≥99.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...