O-Benzylhydroxylamine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

AC10597-0050 AC10597-0250 AC10597-1000
TS10597-0050EA 53 USD
TS10597-0050 TS10597-0250 TS10597-1000
O-Benzylhydroxylamine hydrochloride 99%
O-Benzylhydroxylamine hydrochloride
Formula: C₇H₁₀ClNO
MW: 159.62 g/mol
Melting Pt: 238 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012952
CAS Number: 2687-43-6
EINECS: 220-249-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or flakes
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Water ≤0.1 % (K.F.)
GC ≥91 %
Color scale ≤40 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR