Dimethyl glyoxime disodium salt octahydrate 99%

Supplier: Thermo Scientific Chemicals

AC40834-0010 AC40834-1000 AC40834-2500
TS40834-0010EA 459 USD
TS40834-0010 TS40834-1000 TS40834-2500
Dimethyl glyoxime disodium salt octahydrate 99%
Dimethyl glyoxime disodium salt octahydrate
Formula: C₄H₆N₂Na₂O₂·8H₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150288
CAS Number: 75006-64-3
EINECS: 262-523-2

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Water 43 to 48 % (K.F.)
Solubility (50mg/ml, 50% ethanol) Clear solution
Titration with HClO4 ≥98.5 %
Sensitivity (Test for Ni) Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR