2-Hydroxycyclohexanone dimer 90+%

Supplier: Thermo Scientific Chemicals
Warning

AC39305-0050 AC39305-0250
TS39305-0050EA 35 USD
TS39305-0050 TS39305-0250
2-Hydroxycyclohexanone dimer 90+%
2-Hydroxycyclohexanone dimer
Formula: C₆H₁₀O₂
MW: 228.28 g/mol
Boiling Pt: 83…86 °C (13 mmHg)
Melting Pt: 100…107 °C
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151157
CAS Number: 30282-14-5
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Appearance Off-white Powder
Infrared spectrum Conforms
GC ≥90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR