2-(Chloromethyl)quinoline hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

AC10951-0010 AC10951-0050 AC10951-0250
TS10951-0010EA 31 USD
TS10951-0010 TS10951-0050 TS10951-0250
2-(Chloromethyl)quinoline hydrochloride 97%
2-(Chloromethyl)quinoline hydrochloride
Formula: C₁₀H₉Cl₂N
MW: 214.10 g/mol
Melting Pt: 183…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00012734
CAS Number: 3747-74-8
EINECS: 223-145-3

Order Now

Specification Test Results

Appearance Cream to light yellow Powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %
Solubility (5 % in water) Clear
Titration with HClO4 ≥96.0 %
Iron (Fe) ≤20 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤1000 ppm
Heavy metals (ICP-OES) ≤0.002 %
Sulfur compounds (as SO4) ≤0.2 %
Insoluble matter ≤0.01 % (in dilute HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR