Ammonium iron (III) citrate (16,5 - 18,5% Fe)
Supplier: Thermo Scientific Chemicals
Synonyms:
Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate
AC21184-0010
AC21184-0025
AC21184-0100
AC21184-2500
TS21184-0010EA
233
USD
TS21184-0010
TS21184-0025
TS21184-0100
TS21184-2500
Ammonium iron (III) citrate (16,5 - 18,5% Fe)
Ammonium iron (III) citrate
Formula:
C₆H₅+4y)Fe(x)N(y)O₇ Storage Temperature: Ambient |
MDL Number:
MFCD00013099 CAS Number: 1185-57-5 EINECS: 214-686-6 |
Specification Test Results
Appearance | Brown-yellow to brown Powder |
Infrared spectrum | Conforms |
Assay | 16.5 to 18.5 % (Fe) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...