Lithium ethoxide 1 mol/l in ethanol, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC38017-1000 AC38017-8000
TS38017-1000EA 92 USD
TS38017-1000 TS38017-8000
Lithium ethoxide 1 mol/l in ethanol, AcroSeal™
Lithium ethoxide
Formula: C₂H₅LiO
Density: 0.82 g/cm³
Flash Pt: 9 °C
Storage Temperature: Ambient
MDL Number: MFCD00050493
CAS Number: 2388-07-0
UN: 3274
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to orange Solution
Strength 0.90 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR